This item is now discontinued. Please browse similar items
DWK Life Sciences (Kimble) MOBILE PHASE DELIVERY CAP HPLC
Representative image only.
DWK Life Sciences (Kimble) Mfr # 953913-0000 – Item # RK-02017-35
Find replacements or upgrade your bottle's caps and pouring rings
  • Ensure the bottle cap size is the same as the bottle neck size
  • Polypropylene is autoclavable, PBT and ETFE can withstand dry heat sterilization
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
1 Year Warranty
Key Features
  • Ensure the bottle cap size is the same as the bottle neck size
  • Polypropylene is autoclavable, PBT and ETFE can withstand dry heat sterilization