DWK Life Sciences (Kimble) CHROMAFLEX® Flow Adapter for Standard Columns, 1.0 cm ID x 1/16" OD Tubing

Representative image only.
DWK Life Sciences (Kimble) Mfr # 420836-0000 – Item # RK-34696-54
Optimixe performance of your CHROMAFLEX® columns
  • Recommended when using buffer gradients, during reverse flow applications and for high resolution gravity flow separations

PRODUCT OPTIONS

$543.68CAD / Case of 1

SPECS & DESCRIPTION

Specifications & Description

  • SPE Cartridge Length (mm)150
  • Tubing Outside Diameter (OD) (in)0.002
  • Tubing Inside Diameter (ID) (mm)10
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° C)50
  • DescriptionFlow adapter, for 1.0 cm ID standard columns

Key Features

  • Recommended when using buffer gradients, during reverse flow applications and for high resolution gravity flow separations

More About this Item

Improve chromatogram resolution by delivering the sample and buffer directly to top of gel bed. This protects gel bed from disruption during sample loading and eliminates any dead volume above gel bed. They are ideal for concentration and pH gradients, reverse flow separations, and high-resolution gravity flow. Each includes a 20 µm porosity HDPE bed support, PTFE/propylene O-ring seal, stainless steel adjusting rod, PTFE body and five feet (1.5 m) of FEP PTFE tubing.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat