This item is now discontinued. Please browse similar items
DWK Life Sciences (Kimble) 58502-7 Solvent-Saver Scintillation Vials, HDPE, 7 mL, PE Caps; 1000/PK
Representative image only.
DWK Life Sciences (Kimble) Mfr # 58502-7 – Item # RK-08918-18
Vial trays feature alphanumeric coordinates for fast, easy identification
  • Converted from Type I, 33 expansion borosilicate glass tubing
  • Five trays per case, shrink-wrapped to preserve cleanliness
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)7
  • Tube MaterialHDPE
  • Tube Bottom ShapeFlat
  • Tube ColorNatural
  • Cap Diameter (mm)15
  • Cap MaterialHDPE
  • Description7 mL Solvent-Saver Scintillation Vials, HDPE, PE caps, 1000/cs
Key Features
  • Converted from Type I, 33 expansion borosilicate glass tubing
  • Five trays per case, shrink-wrapped to preserve cleanliness
More About this Item
These scintillation vials are made of made of low potassium borosilicate glass that conforms to ASTM guidelines to deliver optimal background counts and high UV transmission, making them ideal for solvent reduction. Multiple vial and cap configuration options are available for a wide range of applications. All vials include alphanumeric trays to track and organize samples.
Cases of 1000 HDPE vials are shrink-wrapped in trays of 250 vials per container and closures are packaged separately in polyethylene bags.