DWK Life Sciences (Kimble) 20026-250 250 mL class A graduated cylinder
Representative image only.
DWK Life Sciences (Kimble) Mfr # 20026-250 – Item # RK-34527-38
  • Serialized and certified, single metric scale
  • Limited Lifetime Warranty
PRODUCT OPTIONS
$1,163.24CAD / Case of 4
$131.87 - $238.69CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)250
  • Graduation Interval (mL)2
  • Tolerance (mL)±0.80
  • Description250 mL class A graduated cylinder
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Serialized and certified, single metric scale
  • Limited Lifetime Warranty