This item is now discontinued. Please browse similar items
Doran Scale MVPOPT001 Mobility Kit , Casters for Industrial Bench Scales
Representative image only.
Doran Scale Mfr # DMSOPT01 – Item # RK-11930-95
Enhance the versitility of your Doran balance with these accessories
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMobility Kit , Casters for Industrial Bench Scales
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These accesories are compatible with Doran scales.
Optional mobility kit includes a 36" tall column with handles, four 3" diameter locking swivel casters, and indicator mounting plate.
Use with Doran Industrial Bench Scales:
11930-21
11930-22
11930-23
11930-24
11930-26
11930-28
11930-30
11930-32
11930-34
11930-36