This item is now discontinued. Please browse similar items
Diversey Titan™ Tabs Sporicidal Disinfectant Cleaner; Case of 5 x 100 tabs (1 tab dilutes in 32 oz)
Representative image only.
Diversey Mfr # 101102693 – Item # RK-78901-11
Kills Clostridium difficile spores in 4 minutes
  • Broad spectrum disinfectant is effective on bloodborne pathogens, gram positive bacteria, gram negative bacteria, enveloped virus, unenveloped virus, fungus, and spores
  • Convenient tablet form
  • Alternative to bleach
  • Favorable pH is safer for users
SPECS & DESCRIPTION
Specifications & Description
  • FormatTablet
  • Dilution13.1-gram tablet to be diluted in 1 quart of water
  • ApplicationsCleaning and disinfection in hospitals, institutional facilities andmany other industrial uses
  • DescriptionSporicidal Disinfectant Cleaner; Case of 5 x 100 tabs (1 tab dilutes in 32 oz)
Key Features
  • Broad spectrum disinfectant is effective on bloodborne pathogens, gram positive bacteria, gram negative bacteria, enveloped virus, unenveloped virus, fungus, and spores
  • Convenient tablet form
  • Alternative to bleach
  • Favorable pH is safer for users
More About this Item
Titan Tabs can be used for cleaning and disinfection in hospitals, institutional facilities and many other industries. The 13.1-gram tablet must be diluted in 1 quart of water before use. The solution can be applied using spray, mop, wipe, cloth, sponge, brush, trigger sprayer, or mechanical sprayer.