This item is now discontinued. Please browse similar items
Diversey Avert® Sporicidal Disinfectant Cleaner; Case of twelve 32 oz bottles with sprayers
Representative image only.
Diversey Mfr # 100842725 – Item # RK-78901-08
Cleans, disinfects, and deodorizes all in one step
  • Disinfects surfaces in one minute
  • Kills Clostridium difficile spores in four minutes
  • Ready-to-use one-step disinfectant saves you time
  • Effective against bacteria, antibiotic resistant bacteria, viruses, and fungi
SPECS & DESCRIPTION
Specifications & Description
  • FormatSpray bottle
  • Volume (gallons)0.25
  • Volume (Liters)0.95
  • DilutionReady to use
  • ApplicationsClostridium difficile spores, bacteria, antibiotic resistant bacteria, viruses, and fungi sanitizer
  • DescriptionSporicidal Disinfectant Cleaner; Case of Twelve 32 oz Bottles with Sprayers
Key Features
  • Disinfects surfaces in one minute
  • Kills Clostridium difficile spores in four minutes
  • Ready-to-use one-step disinfectant saves you time
  • Effective against bacteria, antibiotic resistant bacteria, viruses, and fungi
More About this Item
Avert® is a ready to use, one-step, all-purpose disinfectant cleaner and deodorizer ideal for eliminating Clostridium difficile (C. diff) spores within four minutes on hard, nonporous surfaces such as glass, glazed porcelain, and stainless steel found in medical and healthcare facilities. Available in a spray solution or premoistened wipes, the bleach-based formula is also effective within a minute against various pathogens including bacteria, antibiotic resistant bacteria, viruses, and fungi.
Meets minimum active ingredient requirements for CDC, SHEA, APIC, and AORN.