This item is now discontinued. Please browse similar items
Digi-Sense Thermocouple, Stranded Wire, Type-J, 20-Gauge, FEP Insulation. 100-ft Spool
Representative image only.
Digi-Sense Item # RK-08541-46
Make your own extension cables
SPECS & DESCRIPTION
Specifications & Description
  • MaterialFiberglass
  • Wire Gauge (Ga)24
  • Inner InsulationFiberglass Braid
  • Outer InsulationFiberglass Wrap
  • Stranded / SolidSolid
  • OverbraidYes
  • Min Temperature (° F)-328
  • Max Temperature (° F)400
  • Min Temperature (° C)-200
  • Max Temperature (° C)204
  • Length (ft)1000
  • DescriptionThermocouple, Stranded Wire, Type-J, 20-Gauge, FEP Insulation. 100-ft Spool
More About this Item
Wire comes in 24-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.