This item is now discontinued. Please browse similar items
Digi-Sense Thermocouple Probe Wire, Type T, 24-Gauge, Glass Braid with SS Overbraid, 1000ft
Representative image only.
Digi-Sense Item # RK-08542-34
Make your own extension cables
SPECS & DESCRIPTION
Specifications & Description
  • MaterialFiberglass
  • Wire Gauge (Ga)20
  • Inner InsulationFiberglass Wrap
  • Outer InsulationFiberglass Wrap
  • Stranded / SolidSolid
  • OverbraidYes
  • Special InformationStainless steel overbraid
  • Min Temperature (° F)-100
  • Max Temperature (° F)900
  • Min Temperature (° C)-73
  • Max Temperature (° C)482
  • Length (ft)1000
  • DescriptionThermocouple Probe Wire, Type T, 24-Gauge, Glass Braid with SS Overbraid, 1000-ft Spool
More About this Item
Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.