This item is now discontinued. Please browse similar items
Digi-Sense Thermocouple Mounting Panel, Horizontal, Standard Connectors; 10 Circuits
Representative image only.
Digi-Sense Item # RK-18527-47
Anodized aluminum construction for full corrosion protection
  • Create easy access to temperature connections in control rooms, test stands, laboratories, and plants
  • Complete your panel assembly with insert panel thermocouple connectors
  • Available in 2, 4, 5 and 6 circuits for vertical face plates and 6, 8, 10 and 12 circuits for horizontal face plates
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Connector TypeStandard connector
  • Special InformationAnodized aluminum face plate/mounting panel
  • Length (in)6 13/16
  • Height (in)1 1/2
  • DescriptionThermocouple Mounting Panel, Horizontal, Standard Connectors; 10 Circuits
  • Warranty6 month
6 Month Warranty
Key Features
  • Create easy access to temperature connections in control rooms, test stands, laboratories, and plants
  • Complete your panel assembly with insert panel thermocouple connectors
  • Available in 2, 4, 5 and 6 circuits for vertical face plates and 6, 8, 10 and 12 circuits for horizontal face plates
  • 6 month warranty
More About this Item
Use to creat easy to access direct temperature connection in control rooms, test stands, laboratories, and plants. Use with insert panel temperature connectors to create a complete panel assembly. Available in 2, 4, 5 & 6 circuits for Vertical Face Plates and 6, 8, 10 & 12 circuits for Horizontal Face Plates.