This item is now discontinued. Please browse similar items
Digi-Sense Temperature Transmitter, Type K, HART protocol, -300 to 300F
Representative image only.
Digi-Sense Item # RK-94710-41
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Control Input ThermocoupleK
  • Output4 to 20 mA
  • Min Temperature (° F)-300
  • Max Temperature (° F)300
  • Accuracy of Span±0.1%
  • Loop Power Supply13 to 40 VDC
  • DescriptionTemperature Transmitter, Type K, HART protocol, -300 to 300F
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Thermocouple and RTD Transmitters