Digi-Sense Temperature Transmitter, Type J, HART protocol, 0 to 500F
Representative image only.
Digi-Sense Item # RK-94710-39
$649.03CAD / Each
$294.53CAD / Each
for an additional
$294.53CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Control Input ThermocoupleJ
  • Output4 to 20 mA
  • Min Temperature (° F)0
  • Max Temperature (° F)500
  • Accuracy of Span±0.1%
  • Loop Power Supply13 to 40 VDC
  • DescriptionTemperature Transmitter, Type J, HART protocol, 0 to 500F
More About this Item
Thermocouple and RTD Transmitters