This item is now discontinued. Please browse similar items
Digi-Sense RTD Jack Panel Set, 3-Jacks for Conduit Box Use; Standard
Representative image only.
Digi-Sense Item # RK-93838-34
Connectors withstand continuous temperature of 350°F (177°C) and maximum temperature of 400°F (204°C)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Connector TypeStandard Connector
  • DescriptionRTD Jack Panel Set, 3-Jacks for Conduit Box Use; Standard
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Feature glass-filled nylon casing with finger grips and polarity indicators; negative terminals are marked with red discs for easy identification. Miniconnectors accept miniature cable clamps and cable up to 20 gauge; standard connectors accept standard cable clamps and cable up to 16 gauge.