This item is now discontinued. Please browse similar items
Digi-Sense Pocket Timer; 5 Minute Markings and Transparent Housing
Representative image only.
Digi-Sense Item # RK-94415-46
Loud—five-second bell rings at zero
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Timing Capacity1 to 60 minutes
  • Count DirectionDown
  • Resolution1 min
  • Alarm5 second ring
  • CaseChemical resistant ABS plastic
  • Depth (in)1
  • Outside Diameter (OD) (in)2 1/2
  • Depth (cm)2.5
  • Outside Diameter (OD) (cm)6.4
  • DescriptionPocket Timer; 5 Minute Markings and Transparent Housing
  • Warranty1 year
1 Year Warranty
Key Features
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • 1 year warranty
More About this Item
Pocket timers are adjustable from 0 to 60 minutes in 1-minute increments. Turning the pointer to the desired countdown setting automatically winds the timers. Indicating pointer always shows exact time remaining.
Model 94415-46 has a 5-minute marking faceplate with transparent housing.