Digi-Sense Pocket Liquid-In-Glass Thermometer, -10 to 110°C (0 to 220°F), Closed Plastic Case, Organic Liquid Fill
Representative image only.
Digi-Sense Item # RK-08008-03
Low-cost way to measure temperature with a compact and durable plastic case
  • Non-mercury blue spirit liquid
  • 5 year warranty
PRODUCT OPTIONS
$25.64CAD / Each
$323.41 - $430.23CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Stay connected — Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")6 19/64
  • Min Temperature (° F)0
  • Min Temperature (° C)-10
  • Max Temperature (° F)220
  • Max Temperature (° C)110
  • Accuracy±2.0°F (±1.0°C)
  • Division (° C)1
  • Division (° F)2
  • Thermometer FillBlue Spirit
  • Dial / Case MaterialClosed Plastic
  • Overall Length (in)6.3
  • Overall Length (mm)160
  • Immersion Depth (mm)160
  • Stem MaterialWhite-Back Glass
  • DescriptionPocket Liquid-In-Glass Thermometer, -10 to 110°C (0 to 220°F), Closed Plastic Case, Organic Liquid Fill
  • Warranty5 years
5 Year Warranty
Key Features
  • Non-mercury blue spirit liquid
  • 5 year warranty
More About this Item
These thermometers are ideal for use in laboratory, biology, chemistry, university, photographic, food/beverage, petrochemical, automotive, wastewater and many other general purpose applications. The protective plastic case protects these thermometers from accidental breakage during use and storage. The section of the case that surrounds the bulb has small openings for circulation. All DURAC Plus® thermometers feature markings that are permanently printed on the glass, blue spirit liquid that has a dark scale contrasts against a white background.