This item is now discontinued. Please browse similar items
Digi-Sense Multifunction AC/DC Voltage Tester with Digital Display & Bar Graph
Representative image only.
Digi-Sense Item # RK-20250-59
Work safely around electricity
  • Test power outlets and switches for correct circuit wiring before resuming power
  • Large, easy-to-read LCD with backlight and bar graph display
  • Stop phantom voltage from interfering with your readings
  • Work in poorly lit or tight areas with built-in flashlight
  • Rugged—IP64 housing with CAT III-1000 V / CAT IV-600 V safety rating
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Max (Volts)690
  • DC Voltage Accuracy0.01
  • DC Current Accuracy0.01
  • AC Min (Volts)6
  • AC Max (Volts)400
  • Ac Voltage Accuracy0.015
  • Resistance Max (KΩ)200
  • Continuity<200kΩ
  • Continuity / Diode Test<1µA
  • Nc Voltage DetectorAutomatic
  • Count1999
  • Current Frequency50/60 Hz ACV
  • DisplayLCD
  • Measurementdouble-pole and contact electrode
  • Safety RatingCAT III 690 V AC/DC
  • DescriptionMultifunction AC/DC Voltage Tester
  • Warranty2 Years
Two year warrantyConformite EuropeenneUL Listed.
Key Features
  • Test power outlets and switches for correct circuit wiring before resuming power
  • Large, easy-to-read LCD with backlight and bar graph display
  • Stop phantom voltage from interfering with your readings
  • Work in poorly lit or tight areas with built-in flashlight
  • Rugged—IP64 housing with CAT III-1000 V / CAT IV-600 V safety rating
  • 2 year warranty
More About this Item
Versatile tester is ideal for your daily electrical measuring tasks—letting you perform AC/DC voltage, low impedance voltage, and continuity testing. Additional functions include full-range polarity detection for DC circuits and rotary field indication to effectively measure phase rotation direction of motors.