This item is now discontinued. Please browse similar items
Digi-Sense Magne-Couple Type K Industrial Probe, with 3" Pigtail
Representative image only.
Digi-Sense Item # RK-93830-05
Ruggedly designed for industrial use, but versatile for use in most applications.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeK
  • Min Temperature (° F)0
  • Max Temperature (° F)1000
  • Min Temperature (° C)17
  • Max Temperature (° C)537
  • Sheath Length (in)72
  • DescriptionMagne-Couple Type K Industrial Probe, with 3\" Pigtail
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Magne-Couple is a thermocouple designed for temporary surface monitoring of any ferrous surface. It attach's to the surface by utilizing a powerful Alnico Magnet with a holding force of 16 lbs, and is capable of withstanding up to 1000 degrees F without degradation. The magnet forces the spring loaded sensing tip into contact with the surface being monitored.