This item is now discontinued. Please browse similar items
Digi-Sense Liquid-in-Glass Thermometer Cup Case; Wood/Brass
Representative image only.
Digi-Sense Item # RK-08081-10
Use for tank thermometer applications
  • 1 year warranty
Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionInstrument Glass Thermometer Wood /Brass Cup Case, Holds 12\" Glass Thermometer
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Wood/Brass cup case is designed with a non-sparking brass cup which holds which holds 20ml of liquid, which meets with both ASTM and API requirements. The thermometer is held in place with spring clips. The hardwood is lacquered to prevent absorption from the sample. Cup case will accommodate up to a 12" (300 mm) glass thermometer. (Thermometer must be ordered separately)