This item is now discontinued. We recommend the following as suggested replacements: RK-08081-12
Digi-Sense Liquid-In-Glass Maximum/Minimum Thermometer; -35 to 50C (-30 to 120F), Organic Liquid Fill
Representative image only.
Digi-Sense Item # RK-90250-00
Reset thermometer with easy-to-use push button
  • For reference only (should not be calibrated)
  • 1 year warranty
Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")7 29/32
  • Min Temperature (° F)-30
  • Min Temperature (° C)-35
  • Max Temperature (° F)120
  • Max Temperature (° C)50
  • Accuracy±1°C at ambient; ±2°C at -30°C tolerance (±2.0°F)
  • Division (° C)1
  • Division (° F)2
  • Thermometer FillRed Spirit
  • Dial / Case MaterialPlastic
  • Overall Length (in)7.9
  • Overall Length (mm)200
  • Stem MaterialGlass
  • DescriptionLiquid-In-Glass Maximum/Minimum Thermometer; -35 to 50C (-30 to 120F), Organic Liquid Fill
  • Warranty1 year
1 Year Warranty
Key Features
  • For reference only (should not be calibrated)
  • 1 year warranty
More About this Item
Efficient thermometer measures both minimum and maximum temperatures in Fahrenheit and Celsius scales. Readings are accurate to ±1 scale division. Bulbs are enclosed in a sturdy plastic case suitable for outdoor or indoor use.