This item is now discontinued. Please browse similar items
Digi-Sense Industrial Thermocouple Element, 4" L, 20gauge, J Type
Representative image only.
Digi-Sense Item # RK-93605-45
Heavy-duty construction for high temperature use
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeJ
  • Min Temperature (° F)-200
  • Max Temperature (° F)2300
  • Min Temperature (° C)-129
  • Max Temperature (° C)1260
  • JunctionGrounded
  • Sheath Length (in)4
  • Sheath Diameter (in)0.032
  • Cable Wire Gauge (Ga)20
  • DescriptionIndustrial Thermocouple Element, 4\" L, 20gauge, J Type
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Elements with in an industrial thermocouple that includes a thermowells that deteriorate over time can be replaced as a more cost effective solution. Each replacement element is made of 20 gauge wire and is encased with ceramic insulators for a greater degree of protection.