This item is now discontinued. Please browse similar items
Digi-Sense General Purpose Liquid-In-Glass Thermometer; -40 to 120F, Total Immersion, Organic Liquid Fill
Representative image only.
Digi-Sense Item # RK-08008-22
Low-cost way to measure temperature quickly and accurately.
  • Non-mercury red spirit liquid
  • 5 year warranty
Stay connected — Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")12
  • Min Temperature (° F)-40
  • Max Temperature (° F)120
  • Accuracy±4.0°F
  • Division (° F)2
  • Thermometer FillRed Spirit
  • Overall Length (in)12
  • Overall Length (mm)300
  • Immersion Depth (mm)300
  • Stem MaterialWhite-Back Glass
  • DescriptionGeneral Purpose Liquid-In-Glass Thermometer; -40 to 120F, Total Immersion, Organic Liquid Fill
  • Warranty5 years
5 Year Warranty
Key Features
  • Non-mercury red spirit liquid
  • 5 year warranty
More About this Item
These thermometers are ideal for use in laboratory, biology, chemistry, university, photographic, food/beverage, petrochemical, automotive, wastewater and many other general purpose applications.
All Durac thermometers feature markings that are permanently printed on the glass, red spirit liquid that has a dark scale contrasts against a white background.