This item is now discontinued. Please browse similar items
Digi-Sense Dual Thermocouple Connector, Mini, Female, Type-T; 1/ea
Representative image only.
Digi-Sense Item # RK-18527-80
Captive cover screw for easy assembly
  • Glass-filled nylon construction with molded barriers to prevent short circuit
  • Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Connector TypeT
  • Special InformationGlass filled nylon body
  • Max Temperature (° F)392
  • Max Temperature (° C)200
  • DescriptionDual Thermocouple Connector, Mini, Female, Type-T; 1/ea
  • Warranty6 month
6 Month Warranty
Key Features
  • Glass-filled nylon construction with molded barriers to prevent short circuit
  • Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue
  • 6 month warranty
More About this Item
Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue.