Digi-Sense Dry Block/Incubator Liquid-In-Glass Thermometer, 18 to 60°C, PFA Safety Coated, 35 mm Immersion, Organic Liquid Fill
Representative image only.
Digi-Sense Item # RK-08006-09
Replace mercury thermometers for a specific application with the same accuracy without the any hazards
  • Non-mercury liquid
  • 5 year warranty
PRODUCT OPTIONS
$111.24CAD / Each
$323.41 - $430.23CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")4 51/64
  • Min Temperature (° C)18
  • Max Temperature (° C)60
  • Accuracy±0.5°C
  • Division (° C)0.5
  • Thermometer FillBlue Spirit
  • Overall Length (in)4.8
  • Overall Length (mm)122
  • Immersion Depth (mm)35
  • Stem MaterialPFA-Coated White-Back Glass
  • DescriptionDry Block/Incubator Liquid-In-Glass Thermometer, 18 to 60°C, PFA Safety Coated, 35 mm Immersion, Organic Liquid Fill
  • Warranty5 years
5 Year Warranty
Key Features
  • Non-mercury liquid
  • 5 year warranty
More About this Item
These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PFA coated for added safety which may cause a slight change in accuracy up to 1 division.
Incubator thermometers - designed with a swell in the glass to hold the thermometer in place.