This item is now discontinued. Please browse similar items
Digi-Sense 2-In-1 Waterproof Infrared Thermometer
Representative image only.
Digi-Sense Item # RK-35625-41
A thermometer for all applications
  • Go/no-go LEDs provide quick check of HACCP food zones
  • IP65 rated waterproof housing
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Distance To Target Size Ratio (D:S)2.5:1
  • EmissivityAdjustable
  • Sighting TypeLED (non-laser)
  • Min Temperature (° F)-67
  • Max Temperature (° F)625
  • Min Temperature (° C)-55
  • Max Temperature (° C)330
  • AccuracyInfrared: ±0.1°F from -27° to 32°F; ±1.0°F from 32° to 149°F; ±1.5% of reading from 149° to 572°F; Contact: ±0.5°F; ±0.9°F above 212° and below 15°F (Infrared: ±0.1°C from -33° to 0°C; ±1.0°C from 0° to 65°C; ±1.5% of reading from 65° to 300°C; Contact: ±0.5°C; ±0.9°C above 100° and below -9°C)
  • Resolution0.5°F (0.2°C)
  • Probe TypeType-K
  • Probe Min Temperature (° F)-55
  • Probe Max Temperature (° F)330
  • Probe Min Temperature (° C)-67
  • Probe Max Temperature (° C)625
  • Display Type1\", 4-digit LCD
  • Low Battery IndicatorYes
  • BatteryAAA Battery (2)
  • Temperature InputYes
  • Description2-In-1 Waterproof Infrared Thermometer
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneIP 65
Key Features
  • Go/no-go LEDs provide quick check of HACCP food zones
  • IP65 rated waterproof housing
  • 1 year warranty
More About this Item
Ultra thin 2-in-1 waterproof contact/noncontact thermometer features a HACCP food safety LED warning. LED’s rapidly indicate if food temperature is within a safe or unsafe range. Green light indicate safe temperatures below 40°F (4°C) and above 140°F (60°C). Red light indicates the HACCP danger zone of 40 to 140°F (4 to 60°C). The built-in stainless steel type-K thermocouple probe swings out for internal temperature checks, then folds in for storage. IR mode provides quick, noncontact surface measurements. Bright white LED pointer illuminates the target measurement area and is ideal for close working distances of 2 to 12”. Product also features adjustable emissivity, Celsius or Fahrenheit selectable, min/max, auto data hold, lock
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.
We now offer an FDA-cleared infrared thermometer.
Our other infrared thermometers are not approved for medical usage and are not FDA-approved.
For help in selecting an infrared thermometer please contact the Technical Support team at Cole-Parmer.