This item is now discontinued. Please browse similar items
Digi-Sense 104A PL512 On/Off Temperature Controller, Type J, –328 to 2192°F, 120V
Representative image only.
Digi-Sense Item # RK-36225-90
A basic on/off controller for processes where temperature stability is less critical
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)32
  • Max Temperature (° F)1832
  • Min Temperature (° C)0
  • Max Temperature (DegreeC)1000
  • Power (VAC)120
  • DescriptionOn/Off Temperature Controller, Type J, –328 to 2192°F, 120V
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This controller is designed to turn the power on and off to the output receptacle based on the set temperature. Equipped with a dial contol that allows you to set and regulate the desired temperature. Features a digital display, selectable temperature scales, and one panel-mointed receptacle. To save bench space, mount the unit to a support pole or lattice