This item is now discontinued. Please browse similar items
Dibafit™ Adapter, 1/4"-28 UNF(M) flat bottom to 3.2 mm ID, acetal; 5/pk
Representative image only.
Diba Mfr # 008NB32-AD5B – Item # RK-21941-41
Use adapters to quickly connect tubing lines or fittings
  • Easily connect soft tubing with a 1/4"-28 UNF(M) connection
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")1 17/64
  • Fitting MaterialAcetal
  • Width (InchShort)25/64
  • Connection TypeBarb x UNF(M)
  • Number Of Ports2
  • Thread size UNF(M) (port 2) (in)1/4\"-28
  • Barb Size / Tubing Id (Port 1) (mm)3.2
  • ColorNatural
  • DescriptionDibafit™ Adapter, 1/4\"-28 UNF(M) flat bottom to 3.2 mm ID, acetal; 5/pk
Key Features
  • Easily connect soft tubing with a 1/4"-28 UNF(M) connection
More About this Item
Designed for use with soft wall tubing, these barbed adapters have a superior barb with a crisp edge to hold the tubing securely in place. Select from acetal or PEEK material of construction.