This item is now discontinued. Please browse similar items
Diba Omnifit® Loop Injection Valve, 6-port
Representative image only.
Diba Mfr # 001127-6LI – Item # RK-21940-24
Designed for low-pressure chromatography systems
  • Choose from single or 6-port valve
SPECS & DESCRIPTION
Specifications & Description
  • Connection Type1/4\"-28 UNF(F)
  • Inlet Port6 ports
  • Housing MaterialPTFE with a PEEK™ casing and PCTFE rotor
  • Max Pressure (PSI)500
  • Max Pressure (bar)33
Key Features
  • Choose from single or 6-port valve
More About this Item
Manual sample injection valve 21939-18 is designed for low-pressure chromatography systems. Easily introduce reproducible sample volumes using the included 0.5 mm, 1.0 mm, and 2.5 mm loops and 5-mL syringe.
Six-port loop injection valve 21940-24 has a "click-stop" mechanism for easy port identification. It is constructed from PTFE with a PEEK™ casing and PCTFE rotor and withstands pressures up to 500 psi (33 bar). Valve accepts 1/4"-28 UNF(M) fittings and features a 0.8 mm bore and M3 mounting holes. Use plug 21940-04 to seal any unused ports.