This item is now discontinued. Please browse similar items
DeWalt DPG54-2D Protector Safety Glasses, Smoke lens
Representative image only.
DeWalt Mfr # DPG54-2D – Item # RK-86497-03
Designed with value in mind from a name you depend on.
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorFrosted Smoke/Yellow
  • Lens ColorSmoke
  • ComplianceANSI Z87.1+ (High Impact)
  • DescriptionProtector Safety Glasses, Smoke lens
Conformite Europeenne
More About this Item
The Protector's streamlined design is perfect protective wear for both men and women. Wraparound polycarbonate lens is distortion free. Rubber-tipped temples help to prevent glasses from falling from workers head.
Compliance: ANSI Z87+ (High Impact)