Bel-Art Desktop Autoclavable HDPE Biohazard Bags, 8.5 x 11"; 100/PK

Representative image only.
Bel-Art Mfr # F13166-0000 – Item # RK-06496-50
  • Perfect for use in fume hoods or personal workstations!

$65.57CAD / Pkg of 100
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • MaterialHDPE
  • Capacity (Gal)0.5
  • ColorOrange and Red
  • Length (in)11
  • Width (in)8.5
  • Length (cm)27.94
  • Width (cm)21.59
  • Thickness (mil)1.25
  • DescriptionDesktop Autoclavable HDPE Biohazard Bags, 8.5 x 11\"; 100/Pk

Key Features

  • Perfect for use in fume hoods or personal workstations!

More About this Item

Place bag on stand (sold separately) for a space-saving, disposal unit on a benchtop. Or, insert hand in an inside out bag to pick up individual pieces. This bright orange-red bag is made of high-molecular weight HDPE and is printed with the biohazard warning symbol. The sterilization indicator patch darkens when autoclaved. Autoclavable at 250°F (121°C).

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat