This item is now discontinued. Please browse similar items
Desco 19641 Test Leads for Surface Resistance Checker, 1 Pair
Representative image only.
Desco Mfr # 19641 – Item # RK-20028-03
Easily determine if a surface is dissipative, conductive, or insulative
  • Ensure compliance to ANSI/ESD S20.20-2007 per ESD TR53
  • Use optional electrodes for resistance measurements of larger surfaces
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTest Leads for Surface Resistance Checker, 1 Pair
Key Features
  • Ensure compliance to ANSI/ESD S20.20-2007 per ESD TR53
  • Use optional electrodes for resistance measurements of larger surfaces
More About this Item
This portable meter is perfect for quick checks of surface resistance for ESD control applications in electronics manufacturing or handling environments. Simply press and hold the test button until the bright LEDs display the reading level. Unit features an automatic test voltage selector allowing the test voltage to switch from 10 V to 100 V if the measured resistance exceeds 1 x 10E5 ohms. Two banana jacks and an electrode toggle switch allow for the connection of two optional external 5-lb electrodes that measure surface resistance point to point, or resistance to equipment ground.