This item is now discontinued. Please browse similar items
Delmhorst HT-4000W/CS Digital Thermohygrometer
Representative image only.
Delmhorst Mfr # HT-4000W/CS – Item # RK-59823-26
Ergonomically designed meter fits right in your hand
  • Versatile—calculates dew point, absolute humidity, and vapor pressure
  • Backlit display makes it easy to read in dimly lit areas or bright sunshine
  • Hold feature freezes reading, ideal when testing hard-to-reach areas
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
1 Year Warranty
Key Features
  • Versatile—calculates dew point, absolute humidity, and vapor pressure
  • Backlit display makes it easy to read in dimly lit areas or bright sunshine
  • Hold feature freezes reading, ideal when testing hard-to-reach areas
  • 1 year warranty
More About this Item
Use this rugged handheld thermohygrometer in a wide range of industrial applications. Meter measures ambient temperature in °F or °C, relative humidity, and calculates dew point, absolute humidity, and vapor pressure. Four directional keys plus a middle key make it easy to navigate around the controls. Unit features low-battery indicators and auto shut-off function to save battery life.