Delmhorst F-2000/PKG F-2000/PKG Digital Hay Moisture Meter Kit

Representative image only.
Delmhorst Mfr # F-2000/PKG – Item # RK-59823-08
Safely store hay bales
  • Ensure correct storage conditions to inhibit fungal rot and avert spontaneous combustion
  • Store up to 100 readings with average and maximum
  • Audible alarm warns you when moisture content exceeds the set point
  • 1 year warranty

PRODUCT OPTIONS

$813.31CAD / Each
Include Electrodes Today!

SPECS & DESCRIPTION

Specifications & Description

  • Measurement TypePin
  • Moisture Range Non-Wood Scale (Min) (%)6
  • Moisture Range Non-Wood Scale (Max) (%)40
  • Display TypeDigital
  • AlarmYes
  • Battery1 9V (included)
  • Height (in)3
  • Width (in)8
  • Depth (in)1 5/8
  • DescriptionF-2000/PKG Digital Hay Moisture Meter Kit
  • Warranty1 year
1 Year Warranty

Includes

F-2000 meter, probe handle with cable, 10" bale prod, short pin prod, and carrying case

Key Features

  • Ensure correct storage conditions to inhibit fungal rot and avert spontaneous combustion
  • Store up to 100 readings with average and maximum
  • Audible alarm warns you when moisture content exceeds the set point
  • 1 year warranty

More About this Item

This moisture meter will help ensure hay is baled and stored at the optimum moisture content to avoid spoilage and prevent the dangerous temperature increase associated with rot which can turn your crop into a fire hazard.
A variety of electrodes are available to customize the meter to your application including three lengths of bale prods, a short pin prod for testing hay in the windrow, and a bale sensor for continuously monitoring the bale chamber from the cab.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat