This item is now discontinued. Please browse similar items
Corning Glass Chromatography Column with Reservoir, 250 mL
Representative image only.
Corning Mfr # LG4564T-104 – Item # RK-34695-56
Ideal for adsorption, gel, permeation, and ion exchange techniques
  • Perfect for flash chromatography applications
  • Easily control the flow rate
SPECS & DESCRIPTION
Specifications & Description
  • SPE Cartridge Length (mm)150
  • Total Volume Capacity (mL)250
  • DescriptionGlass Chromatography Column with Reservoir, 250 mL
Key Features
  • Perfect for flash chromatography applications
  • Easily control the flow rate
More About this Item
LabGlass Chromatography Columns with attached reservoirs are perfect for flash chromatography applications. Columns include a PTFE plug to control flowrate. Standard joint size is TS 24/40. Also available with coarse fritted TS 35/25 joint size; call our Application Specialists for details.