This item is now discontinued. Please browse similar items
Corning Gosselin FBAG-03 Blender Bag with Full Filter, polyethylene, 400 mL; 1500/cs; 3 boxes x 20 bags of 25
Representative image only.
Corning Gosselin Mfr # FBAG-03 – Item # RK-34551-44
PE Sample blender bags with optional side filter
  • Use as universal bags for any of your applications
  • Without or with a non woven filter for reduced particles
  • Each filter bag has a printed lot number
  • Smart choice for suspension sampling with pipets
  • Smart choice for homogenous suspension
  • Sterile
SPECS & DESCRIPTION
Specifications & Description
  • Bag TypePE
  • Bag Capacity (mL)400
  • Height (in)11 51/64
  • ColorNatural
  • Description Blender Bag with Lateral Filter, polyethylene, 400 mL, without symbol; 1500/pk
Key Features
  • Use as universal bags for any of your applications
  • Without or with a non woven filter for reduced particles
  • Each filter bag has a printed lot number
  • Smart choice for suspension sampling with pipets
  • Smart choice for homogenous suspension
  • Sterile
More About this Item
Use Gosselin™ blender bags with the Gosselin Adjustable Sample Blender (catalog MM-34551-08) or as universal bags. The sterile PE material sample bags are available in 80 mL and 400 mL sizes. The sample bags are available without and with a side filter to reduce particles from a primary dilution stage. The bag design improves the pipetting process after homogenization. The closure clip is sold separately with catalog number MM-34551-65.