Corning Gosselin Cell Spreader, Sterile, Polystyrene, L-Shaped, White, 148 mm L; 6000/Cs
Representative image only.
Corning Gosselin Mfr # ETAR-05 – Item # RK-34551-46
Safe, efficient sample plating
  • Sterile polystyrene (PS)—no need to preclean
  • Eliminate the risk of cross-contamination
  • Available in L shape and T shape to cover all uses
  • Conveniently packaged in resealable bags
  • Packed 50/Bag, 1200/Box, 6000/Case
PRODUCT OPTIONS
$1,856.47CAD / Case of 6000
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Overall Length (mm)148
  • Handle TypeL-Shaped
  • ColorWhite
  • MaterialPolystyrene
  • DescriptionCorning Gosselin Cell Spreader, Sterile, Polystyrene, L-Shaped, White, 148 mm L; 6000/Cs
Sterile
Key Features
  • Sterile polystyrene (PS)—no need to preclean
  • Eliminate the risk of cross-contamination
  • Available in L shape and T shape to cover all uses
  • Conveniently packaged in resealable bags
  • Packed 50/Bag, 1200/Box, 6000/Case
More About this Item
These sterile disposable cell spreaders are ideal for evenly spreading bacterial cultures while reducing the risk of cross contamination. Spreaders are color coded for easy identification.
L-shaped spreader quickly swirls the culture sample around the petri dish.
T-shaped spreader offers more precise maneuverability for detailed spreading protocols.