This item is now discontinued. Please browse similar items
Corning CoolRack® LV Cooling Block, 12 x 13 or 16 mm Dia. Tubes, Lightweight; Purple
Representative image only.
Corning Mfr # 432069 – Item # RK-04392-69
Cool, freeze, heat, or thaw samples uniformly
  • Place blocks directly into a temperature source from -196°C to over +100°C!
  • Obtain temperature uniformity of ±0.1°C across tubes and plates
  • Quickly reach bath temps—thermoconductive metal construction
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Tubes Held12
  • ColorGray
  • Capacity12 x 13/16 mm diameter tubes
  • DescriptionCooling Block, 12 x 13 or 16 mm Dia. Tubes, Lightweight; Gray
  • Warranty1 year
1 Year Warranty
Key Features
  • Place blocks directly into a temperature source from -196°C to over +100°C!
  • Obtain temperature uniformity of ±0.1°C across tubes and plates
  • Quickly reach bath temps—thermoconductive metal construction
  • 1 year warranty
More About this Item
These aluminum blocks provide better temperature uniformity than placing samples directly into a bath. Aluminum blocks may be autoclaved, high-heat sterilized, or chemically treated to decrease cross-contamination concerns. Safe for use in ice, dry ice, alcohol, liquid nitrogen, water, oil, and other temperature regulation baths. Use in traditional ice buckets and pans, or use with a Corning CoolBox™ for an ice-free option.