Corning Combination-Angled Sidearm Fittings for 1 L Culture Flasks, 1/4" Inlet

Representative image only.
Corning Mfr # 4519-162 – Item # RK-01837-44
Optimize performance of your spinner flasks
  • Compatible with the Corning® reusable spinner flasks
  • Fits 1 L angled sidearm spinner flask
  • Autoclavable

PRODUCT OPTIONS

$604.09CAD / Each
Include Reusable Spinner Flasks Today!
Corning® Reusable Spinner Flasks
$1,134.51 - $2,077.48CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPET insert, fluorocarbon O-Ring and polypropylene sealing cap
  • Diameter (in)0.25
  • Capacity (mL)1000
  • DescriptionCombination-angled sidearm fittings for 1L Culture Flasks

Key Features

  • Compatible with the Corning® reusable spinner flasks
  • Fits 1 L angled sidearm spinner flask
  • Autoclavable

More About this Item

Choose from a wide range of accessories and replacement parts suitable for use with the Corning® reusable spinner flasks.

Combination Fittings include one tube which is bent and another shorter tube which is straight. The
straight tube is approximately 6 inches long and is intended to be located so that its open end is above
the level of liquid in the flask.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat