Corning Dual-Angled Sidearm Fittings for 1 L Culture Flasks, 1/4" Inlet

Representative image only.
Corning Mfr # 4519-154 – Item # RK-01837-40
Optimize performance of your spinner flasks
  • Compatible with the Corning® reusable spinner flasks
  • Fits 1 L angled sidearm spinner flask
  • Autoclavable

PRODUCT OPTIONS

$604.09CAD / Each
Include Reusable Spinner Flasks Today!
Corning® Reusable Spinner Flasks
$1,134.51 - $2,077.48CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPET insert, fluorocarbon O-Ring and polypropylene sealing cap
  • Diameter (in)0.25
  • Capacity (mL)1000
  • DescriptionDual-angled sidearm fittings for 1L Culture Flasks

Key Features

  • Compatible with the Corning® reusable spinner flasks
  • Fits 1 L angled sidearm spinner flask
  • Autoclavable

More About this Item

Choose from a wide range of accessories and replacement parts suitable for use with the Corning® reusable spinner flasks.

Dual Fittings include two tubes which are both bent so that once past the angled sidearm, the
tubes terminate vertically, are below the liquid level in the flask, in the region between the impeller
assembly and the side wall of the flask.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat