Corning Roller Bottle, Sterile, Polystyrene, Tissue Culture Treated, Easy Grip Cap, 850 cm2; 40/Cs
Representative image only.
Corning Mfr # 430849 – Item # RK-01836-01
Seamless, one-piece construction reduces risk of breakage
  • Cell culture treated for optimal cell attachment
  • Individually printed with lot numbers for easy identification and traceability
  • Easy grip caps have smooth edges and large knurls for easy opening—just twist a half turn
  • Bottles have black printed graduations
  • 2 year warranty
PRODUCT OPTIONS
$891.40CAD / Case of 40
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene Bottle and HDPE Cap
  • ClosureEasy Grip Cap
  • Height (mm)271
  • Diameter (mm)117
  • Culture Area (cm²)850
  • SterileYes
  • Capacity (mL)255
  • DescriptionRoller Bottle, Sterile, Polystyrene, Tissue Culture Treated, Easy Grip Cap, 850 cm2; 40/Cs
  • Warranty2 year
Two year warranty
Key Features
  • Cell culture treated for optimal cell attachment
  • Individually printed with lot numbers for easy identification and traceability
  • Easy grip caps have smooth edges and large knurls for easy opening—just twist a half turn
  • Bottles have black printed graduations
  • 2 year warranty
More About this Item
Optically-clear polystyrene roller bottles are sterilized by gamma radiation and certified nonpyrogenic. Bottles are tissue culture treated for optimal cell attachment. Bottles are constructed from durable polystyrene and caps are made of HDPE. Models 01836-05 and -06 feature a ribbed design that doubles the growth area without increasing bottle size. Two window areas provide microscopic viewing as well as pouring surfaces.
Choose from three cap type options. The plug seal provides a liquid-tight seal and the Easy grip cap is designed for easy opening. The Easy grip vent cap adds a 0.2 μm nonwetting (hydrophobic) membrane sealed to the cap.