This item is now discontinued. Please browse similar items
Corning 2158-250 Concentrator with Springs, 250 Ml
Representative image only.
Corning Mfr # 2158-250 – Item # RK-34530-04
Efficiently boil away pestistides for accurate testing
  • Held together using glass hooks and two 1 1/4 inch stainless steel springs
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeConcentrator
  • Range250 mL
  • DescriptionConcentrator with Springs, 250 Ml
Key Features
  • Held together using glass hooks and two 1 1/4 inch stainless steel springs
More About this Item
Kuderna-Danish Concentrators boil away samples to allow testing for pesticides or other pollutants prior to instrumental analysis. Include three-ball Snyder column with venting dimples and graduated collection tube. Available with springs mounted on collection tube.