This item is now discontinued. Please browse similar items
Corning 2157-500 Kuderna-Danish Concentrator without springs, 500 mL capacity, 615mm H
Representative image only.
Corning Mfr # 2157-500 – Item # RK-34530-01
Efficiently boil away pestistides for accurate testing
  • Keck clamps instead of spring hooks
  • All columns have venting dimples
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)500
  • Height (in)24.21
  • DescriptionKuderna-Danish Concentrator without springs, 500 mL capacity, 615mm H
Key Features
  • Keck clamps instead of spring hooks
  • All columns have venting dimples
More About this Item
Kuderna-Danish Concentrators boil away samples to allow testing for pesticides or other pollutants prior to instrumental analysis. Include three-ball Snyder column with venting dimples and graduated collection tube. Available without springs mounted on collection tube.