This item is now discontinued. Please browse similar items
Corning 2157-1L Kuderna-Danish Concentrator without springs, 1L capacity, 720mm H
Representative image only.
Corning Mfr # 2157-1L – Item # RK-34530-03
Efficiently boil away pestistides for accurate testing
  • Keck clamps instead of spring hooks
  • All columns have venting dimples
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)1000
  • Height (in)28.35
  • DescriptionKuderna-Danish Concentrator without springs, 1L capacity, 720mm H
Key Features
  • Keck clamps instead of spring hooks
  • All columns have venting dimples
More About this Item
Kuderna-Danish Concentrators boil away samples to allow testing for pesticides or other pollutants prior to instrumental analysis. Include three-ball Snyder column with venting dimples and graduated collection tube. Available without springs mounted on collection tube.