This item is now discontinued. View the direct replacement: RK-63100-71 or browse similar items

CoorsTek 66325 Porcelain Mortar, 750 mL, pack of 1

Representative image only.
CoorsTek Mfr # 66325 – Item # RK-17820-10

SPECS & DESCRIPTION

Specifications & Description

  • Mortars Capacity (mL)750
  • Mortars Top OD (mm)163
  • Mortars Height (mm)110
  • DescriptionPorcelain Mortar, 750 mL, pack of 1

More About this Item

Mortars are glazed all over except for grinding surfaces and the outside bottom. Features include lip and pour spout. Mortar and pestle are sold as separate pieces. Autoclavable.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat