This item is now discontinued. Please browse similar items
CoorsTek 66015 Permanent Ceramic Marking Ink, 15 mL
Representative image only.
CoorsTek Mfr # 60015 – Item # RK-17829-00
Permanently mark porcelain and ceramic labware
  • Easy to apply with pen or brush Latex free
SPECS & DESCRIPTION
Specifications & Description
  • Pen ColorBlack
  • DescriptionPermanent Ceramic Marking Ink, 15 mL
Key Features
  • Easy to apply with pen or brush Latex free
More About this Item
Use to permanently mark laboratory porcelain and ceramicware. You can apply with pen or brush. Markings become permanent when heated.