This item is now discontinued. View the direct replacement: RK-17829-63 or browse similar items
CoorsTek 66320 Ceramic/Porcelain Pestle, 180 mm Length; 1/Pk
Representative image only.
CoorsTek Mfr # 66320 – Item # RK-17821-06
SPECS & DESCRIPTION
Specifications & Description
  • Pestles Length (mm)180
  • DescriptionCeramic/Porcelain Pestle, 180 mm Length; 1/Pk
More About this Item
Pestles are glazed all over except for grinding surfaces. Features an oversized pestle for comfort. Mortar and pestle are sold as separate pieces. Autoclavable.