This item is now discontinued. We recommend the following as suggested replacements: RK-17829-51
CoorsTek 66298 Hirsch Funnels, Ceramic, 20 mL, 1/pk
Representative image only.
CoorsTek Mfr # 66298 – Item # RK-17819-02
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCeramic
  • Disc Diameter (mm)55
  • Filter Paper Diameter (mm)15
  • DescriptionHirsch Funnels, Ceramic, 20 mL, 1/pk
More About this Item
Fixed perforated funnel plate accepts standard filters. All are glazed inside and out except for the rim. Autoclavable.