This item is now discontinued. Please browse similar items
Control Products AD148-12/200F Temperature SwitchesTemperature Trip Point; 200F
Representative image only.
Control Products Mfr # AD148-12/200F – Item # RK-93880-58
Use to control system temperatures or to act as a final safety shutoff
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)200
  • Max Temperature (° F)200
  • Length (in)1 1/4
  • Diameter (in)11/16
  • DescriptionTemperature SwitchesTemperature Trip Point; 200F
More About this Item
Designed for use in rigorous industrial or laboratory condition to control overheating of air compressors, distilling machines, and other equipment. A bimetal snap disc reverses from convex to concave upon reaching temperature trip point, which activates a SPDT microswitch.
The three-wire design allows you to wire SPDT switch for either normally open (NO) or normally closed (NC) operation. Simple to install—switches require very little wiring and don't need shock mounts. Compact enough to easily thread into any 1/2" NPT opening. Stainless steel housing.