This item is now discontinued. Please browse similar items
ABB RVDA6V25 Continental 1-Phase Solid-State Contactors; 660VAC/25A/DC Logic
Representative image only.
ABB Mfr # SSM1A430BD – Item # RK-94469-52
SPECS & DESCRIPTION
Specifications & Description
  • Load Rating660 VAC
  • Control Voltage4 to 32 VDC
  • Max Current (amps)25
  • Input LogicOn: 4 VDC (at 5.4 mA) to 32 VDC (at 10 mA)
  • DescriptionContinental 1-Phase Solid-State Contactors; 660VAC/25A/DC Logic
More About this Item
FEATURES
  • Superior Surge Survival Technology
  • 25 or 40 amp ratings
  • 575 or 660V max
  • Integrated heatsink
  • LED input indicator
  • Direct copper bonded SCRs
  • Mounts on DIN rail or panel
  • Meets En60947-4-3 and EN55011
  • UL recognized, CSA listed
  • Zero voltage turn-on (CIRVMA units)