This item is now discontinued. Please browse similar items

Compound vials, 14.75 x 22 mm, 1.25 mL, clear glass, 144/cs

Representative image only.
Mfr # GLC-08764 – Item # RK-03757-16
Ideal for storage of small amounts of compounds such as reference standards
  • Safely and securely store samples

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)1.25
  • Tube MaterialBorosilicate Glass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Height (in)7/8
  • Outside Diameter (in)37/64
  • Height (mm)22
  • Outside Diameter (mm)14.75
  • DescriptionCompound vials, 14.75 x 22 mm, 1.25 mL, clear glass, 144/cs

Key Features

  • Safely and securely store samples

More About this Item

These borosilicate glass vials are available in clear or amber glass. Vials are low profile for greater stability and wide mouth for ease of sampling. Cases contain a shrink-wrapped tray vials including caps.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat