COMARK RAYMTFSU Infrared (IR) Thermometer for Food
Representative image only.
COMARK Mfr # COPMTFSU – Item # RK-39641-20
Thermometer has been specifically designed for use in food applications
  • Noncontact thermometer eliminates the risk of cross-contamination
  • Laser sighting pinpoints the center fo the measurement area
  • Fast, accurate readings allow you to take readings more frequently
  • 1 year warranty
$196.40CAD / Each
$263.74 - $371.29CAD / Each
for an additional
$263.74CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Distance To Target Size Ratio (D:S)4:1
  • EmissivityFixed
  • Sighting TypeSingle-point laser
  • Laser ClassClass II
  • Min Temperature (° F)-25
  • Max Temperature (° F)400
  • Accuracy±2.0°F (±1.0°C)
  • Resolution
  • Response time<500 milliseconds
  • Display TypeLCD
  • Battery1 9V (not included)
  • DescriptionRAYMTFSU Infrared (IR) Thermometer for Food
  • Warranty1 year
1 Year Warranty
Key Features
  • Noncontact thermometer eliminates the risk of cross-contamination
  • Laser sighting pinpoints the center fo the measurement area
  • Fast, accurate readings allow you to take readings more frequently
  • 1 year warranty
More About this Item
Measure surfaces within ± 2°F of the critical temperature zone of 32 to 150°F- the range where harmful bacteria grows most rapidly. Scan the surfaces of numerous foods quickly and often- an ideal way to monitor food temperatures for your daily HACCP routine,
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.
We now offer an FDA-cleared infrared thermometer.
Our other infrared thermometers are not approved for medical usage and are not FDA-approved.
For help in selecting an infrared thermometer please contact the Technical Support team at Cole-Parmer.